Check Order Status
Borosilicate Glass Round Bottom Test Tubes
Actual product may vary
Fisherbrand
Borosilicate Glass Round Bottom Test Tubes
Versatile, round-bottom test tubes constructed of borosilicate. Pyrex™ Borosilicate Glass Round Bottom Test Tubes are supplied in packages of 100.
Have Questions?
Catalog Number
Diameter (Metric)
FKS12024617
20 mm
Catalog number
FKS12024617
Price (SGD)
454.00
PK
Add to cart
Diameter (Metric):
20 mm
Price (SGD)
454.00
PK
Add to cart
Borosilicate glass construction
Supplied in packs of 100
Specifications
Capacity (Metric)
40 mL
Length (Metric)
200 mm
Material
Borosilicate Glass
Quantity
100 Pack
Diameter (Metric)
20 mm