1x1 image pixel for data collection
Turbidity Standards for Orion™ AQ3010 Turbidity Meter
Product Image
Fisherbrand
Turbidity Standards for Orion™ AQ3010 Turbidity Meter
Replacement Turbidity Standards for the Orion AQ3010 Turbidity meter.
Have Questions?
Catalog NumberDescription
FKS12013273Replacement calibration standards kit, includes 800, 100, 20.0 and 0.02 NTU standards in plastic bottles
Catalog number FKS12013273
Price (SGD)
1,747.00
PK
Add to cart
Description:
Replacement calibration standards kit, includes 800, 100, 20.0 and 0.02 NTU standards in plastic bottles
Price (SGD)
1,747.00
PK
Add to cart
Turbidity Standards are for use with the Orion™ AQ3010 Turbidity Meter (Cat. No. AQ3010). These replacement calibration standards kit are polymer-based primary turbidity standards and includes 800, 100, 20 and 0.02 NTU in 60mL plastic bottles.
  • For use with: Orion AQUAfast AQ3010
  • Standards Set has 60 mL bottle each of 0.02, 20.0, 100, and 800 NTU 100
Specifications
TypeCalibration Standards
For Use With (Equipment)Orion AQ3010 Turbidity Meter
DescriptionReplacement calibration standards kit, includes 800, 100, 20.0 and 0.02 NTU standards in plastic bottles
Includes800, 100, 20.0 and 0.02 NTU standards in 60mL bottles
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link