1x1 image pixel for data collection
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Actual product may vary
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Fisherbrand
Orion™ Carbon Dioxide Combination Ion Selective Electrode
Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly, accurately and economically.
Have Questions?
Catalog NumberConnector Types
FKS11996069BNC (Waterproof)
Catalog number FKS11996069
Price (SGD)
4,355.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
4,355.00
EA
Add to cart
Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately with the Thermo Scientific™ Orion™ 9502BNWP Carbon Dioxide Electrode. 
Features:
  • It features a gas-permeable membrane technology for use with loose membranes and O-rings.
Solutions required:
  • Standard
    • 950206 0.1 M NaHCO3 Carbon Dioxide standard, 475 mL
    • 950207 1000 ppm as CaCO3 Carbon Dioxide standard, 475 mL
  • Carbon dioxide buffer solution
    • 950210 Carbon Dioxide Ionic Strength Adjuster (ISA), 475 mL
  • Electrode fill solution
    • 950202 Carbon Dioxide electrode fill solution, 60 mL
  • Storage solution
    • 941706 0.1 M NaCl Chloride standard, 475 mL
Items included with:
  • 950202 Carbon Dioxide electrode fill solution, 60 mL 1 bottle
  • 950204 4 membranes with 1 O-ring
Warranty period:
  • 12 months from date of purchase
Specifications
Connector TypesBNC (Waterproof)
DescriptionCarbon Dioxide Electrode
TypeCarbon Dioxide Electrode
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link