Glass Graduated Class B Type 1 Pipettes
Glass Graduated Class B Type 1 Pipettes
Actual product may vary
Fisherbrand
Glass Graduated Class B Type 1 Pipettes
Pipet liquids securely. Fisherbrand™ graduated class B type 1 pipets are made from soda lime glass. Type 1 pipets have the zero at the top.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsTolerance
FKS119921681 mL0.05 mL±0.05 mL
FKS1192217825 mL0.1 mL±0.10 mL
FKS1191217810 mL0.2 mL±0.20 mL
FKS119021785 mL0.2 mL±0.20 mL
Catalog number FKS11992168
Price (SGD)
37.00
PK
Add to cart
Volume (Metric):
1 mL
Graduations:
0.05 mL
Tolerance:
±0.05 mL
Price (SGD)
37.00
PK
Add to cart
  • Soda lime Glass
  • Blue ceramic graduations
  • Type 1 Pipet
  • Class B
Specifications
Certifications/ComplianceClass B, BS 700 ISO 835
ClassB
MaterialSoda Lime Glass
GraduatedYes
Graduations0.05 mL
Quantity5 Pack
Tolerance±0.05 mL
Volume (Metric)1 mL
TypePipet
Unit SizePK