Check Order Status
Whatman™ Mixed Cellulose Ester Membrane Filters
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Whatman™ Mixed Cellulose Ester Membrane Filters
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
Have Questions?
Contact Us
Catalog Number
Pore Size
Diameter (Metric)
Color
Membrane Type
FKS11982006
0.2 μm
47 mm
White
ME24
Catalog number
FKS11982006
Price (SGD)
2,507.00
PK
Estimated availability date
01-Oct-2025
Add to cart
Pore Size:
0.2 μm
Diameter (Metric):
47 mm
Color:
White
Membrane Type:
ME24
Price (SGD)
2,507.00
PK
Add to cart
Sterile options available for critical applications
Contrast for easier particle detection
Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
Higher dirt loading capacity
Biologically inert with good thermal stability
No surfactants to contaminate samples
Uniform microporous structure of membrane gives high flow rates
Specifications
Autoclavable
Autoclavable
Color
White
Material
MCE
Pore Size
0.2 μm
Sterility
Sterile
Grid Color
Black
Hydrophobicity
Hydrophilic
Membrane Type
ME24
No. per Pack
400
Protein Binding Capacity
Medium
Size (Metric) Grid
0.31cm
Surface Type
Grid
Temperature (English) Operating
266°F
Temperature (Metric) Operating
130°C
Type
Membrane Filter
Diameter (Metric)
47 mm
Unit Size
PK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link