Pyrex™ 60mm Diameter Glass Büchner Filter Funnel
Pyrex™ 60mm Diameter Glass Büchner Filter Funnel
Actual product may vary
Fisherbrand
Pyrex™ 60mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
Have Questions?
Catalog NumberPorosityPore Size
FKS11981025316 to 40μm
Catalog number FKS11981025
Price (SGD)
1,699.00
PK
Add to cart
Porosity:
3
Pore Size:
16 to 40μm
Price (SGD)
1,699.00
PK
Add to cart
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
  • Choice of disc diameters and porosities
Specifications
Capacity (Metric)125 mL
Diameter (Metric) Stem13mm
MaterialBorosilicate Glass, Borosilicate Glass
Height (Metric)180 mm
For Use With (Application)For convenient filtration of solutions
Diameter (Metric) Disc60mm
IncludesIntegrated filter plate sintered Glass disc
Length (Metric) Stem100mm
Porosity3
Pore Size16 to 40μm
Product TypeBuchner Funnel
Unit SizePK