Polypropylene Micro Tube Rack
Polypropylene Micro Tube Rack
Actual product may vary
Fisherbrand
Polypropylene Micro Tube Rack
Store 1.5 to 2.0 mL microtubes in color-coded racks. Polypropylene Micro Tube Rack stores up to 80 screw-top vials for benchtop use, storage or transport of samples. Color-coded racks feature molded-in coordinates and handles for easy handling. Autoclavable and suitable for freezing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11978074Green
FKS11728084Assorted
FKS11720344Natural
Catalog number FKS11978074
Price (SGD)
86.00
PK
Add to cart
Color:
Green
Price (SGD)
86.00
PK
Add to cart
Popular fraction-collector-plate style rack
  • Holds 80 screw-cap microtubes or cryogenic vials in 1 mm diameter wells
  • Stores 1.5 to 2.0 mL tubes in compact arrangement (5 x 16 array)
  • Color coded to easily identify specific laboratory procedures or individual technicians
  • Molded-in coordinates for easy sample organization and location
  • Constructed of sturdy polypropylene with molded handles for easy handling
  • Autoclavable and suitable for freezing
Specifications
Product TypeMicrotube Rack
Closure IncludedNo
Array5 x 16
AutoclavableAutoclavable
ColorGreen
HoldsMicrotubes (1.5 to 2 mL)
MaterialPolypropylene
Positions80
ShapeRectangular
ShelvesNone
StackableYes
SterilityNon-sterile
For Use With (Application)Ideal for Freezer Storage
For Use With (Equipment)Microcentrifuge Tube
No. per Pack5 Pk.
Quantity5 Pk.
Length (Metric)225 mm
Length (English)8.9 in.
Width (Metric)67 mm
Width (English)2.6 in.
Height (Metric)28 mm
Height (English)1.1 in.
Diameter (Metric) Well11 mm
Unit SizePK