Foil Roll
Foil Roll
Actual product may vary
Fisherbrand
Foil Roll
Cut foil to any size length. Foil Roll can be used with the Mini Bin Foil Dispenser or on its own.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (Metric)
FKS11977994153 mm
FKS11967994102 mm
Catalog number FKS11977994
Price (SGD)
105.00
EA
Add to cart
Width (Metric):
153 mm
Price (SGD)
105.00
EA
Add to cart
  • Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and Petri dishes.
  • Ideal for autocalving, freezing, incubating, protecting labware, lining, sealing, and general storage.
  • 152.4 m of foil available in two widths: 102 mm or 153 mm
  • Disposable
  • Designed to fit the Mini Bin Foil Dispenser.
Specifications
ApplicationsAutoclaving, freezing, incubating, protecting labware, lining, sealing, and general storage
Length (English)500 ft.
ColorMetallic
Product TypeAluminum Foil
Width (English)6 in.
Width (Metric)153 mm
Length (Metric)152.4 m
MaterialAluminum
Thickness15 μm
For Use With (Equipment)Mini Bin Foil Dispenser
Unit SizeEA