SureOne™ Aerosol Barrier Pipette Tips
SureOne™ Aerosol Barrier Pipette Tips
Actual product may vary
SureOne™ Aerosol Barrier Pipette Tips
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleColor
FKS119777140.1 to 20 μLExtended Length, Micro Point, Low RetentionClear
FIS02-707-4390.1 to 10 μLMicro PointNatural
FIS02-707-4420.1 to 10 μLMicro Point, Extended LengthNatural
FIS02-707-4700.1 to 20 μLExtended Length, Micro Point, Low RetentionClear
FIS02-707-43110 to 100 μLBeveledNatural
FIS02-707-430
also known as FIS#02-707-430
20 to 200 μLMicro PointNatural
FKS1196346620 to 200 μLMicro PointNatural
Catalog number FKS11977714
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
0.1 to 20 μL
Tip Style:
Extended Length, Micro Point, Low Retention
Color:
Clear
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
Specifications
Volume (Metric)0.1 to 20 μL
FilteredFiltered
SterilitySterile
ColorClear
AutoclavableNot Autoclavable
Length (Metric)46 mm
FormatHinged Rack
Tip StyleExtended Length, Micro Point, Low Retention
DisposableReusable
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineSureOne
Graduations2 μL, 5 μL, 10 μL
Unit SizePK