Check Order Status
SuperFrost Ultra Plus™ GOLD Adhesion Slides
Fisherbrand
SuperFrost Ultra Plus™ GOLD Adhesion Slides
Epredia™TM SuperFrost Plus™ Gold slides are recommended for hard-to-hold tissue samples like bone, brain, breast and skin. They are ideal for special stains, immunocytochemical and in-situ DNA hybridization on these tissue types.
Have Questions?
Catalog number
FKS11976299
Price (SGD)
258.00
PK
Add to cart
Price (SGD)
258.00
PK
Add to cart
No time wasted on slide-coating proceduress
No money wasted on expensive adhesives
Reliable and uniform coating
Minimal sample loss
Reliable and safe adhesive action
Specifications
Length (Metric)
75 mm
Width (Metric)
25 mm
Color
White