Polystat™ Immersion Circulator
Polystat™ Immersion Circulator
Actual product may vary
Polystat™ Immersion Circulator
Polystat™ Immersion Circulator
Polystat™ Immersion Circulator
Fisherbrand
Polystat™ Immersion Circulator
Includes fixing clip. Fisherbrand™ Polystat™ Immersion Circulator is DIN 12876-1 certified.
Have Questions?
Catalog NumberTemperature RangeFlow RateIncludesTemperature Stability
FKS1197022820°C to 150°C15 L/min.Stainless Steel lid±0.03°C
Catalog number FKS11970228
Price (SGD)
-
Temperature Range:
20°C to 150°C
Flow Rate:
15 L/min.
Includes:
Stainless Steel lid
Temperature Stability:
±0.03°C
  • Temperature range of 20 to 150°C
  • Electrical requirements: 230V, 50Hz, 9A
  • Includes wall fixing clip
  • Flow Rate of 15L/min
  • Weight: 4.1kg
  • Temperature Stability of ±0.03°C
Specifications
Capacity (Metric)5 L
Certifications/ComplianceClassification III (NFL) according to DIN 12876-1
Flow Rate15 L/min.
Temperature Range20°C to 150°C
TypeImmersion Circulator
Heating Capacity2 kW
IncludesStainless Steel lid
ModelPolystat 36
Temperature Stability±0.03°C
Width (Metric)13 cm
Height (Metric)33 cm
Depth (Metric) Interior8 to 14.5 cm
Weight (Metric)4.1 kg
Electrical Requirements230 V 50 Hz 9 A
Voltage230 V
Amperage9 A
Frequency50 Hz
Unit SizeEA