Pyrex™ Borosilicate Glass Reagent Bottles
Pyrex™ Borosilicate Glass Reagent Bottles
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Reagent Bottles
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1196159850 mL
FKS1194032625 mL
FKS10105044100 mL
FKS10411235250 mL
FKS10786885500 mL
Catalog number FKS11961598
Price (SGD)
258.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
258.00
PK
Add to cart
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
  • Complies with ISO 4796-1
  • Manufactured from Pyrex™ borosilicate glass
  • Excellent resistance to chemical attack and high temperatures
  • Supplied with PBT screw caps that have a PTFE faced sealing disc
  • Autoclavable
  • White graduations and marking spot
Specifications
Capacity (Metric)50 mL
For Use With (Application)Ideal for storage of reagents, analytical standards and culture media etc
MaterialBorosilicate Glass
No. per Pack10/PK
Closure ColorBlack
Certifications/ComplianceISO 4796-1
Closure IncludedYes
AutoclavableAutoclavable
GraduatedYes
Height (Metric)80 mm
Closure LinerPTFE
Closure MaterialPhenolic
Closure Size25 mm
Diameter (Metric)43 mm
Closure TypeScrew Cap
Unit SizePK