Microplate Reader Verification Plates
Microplate Reader Verification Plates
Actual product may vary
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Microplate Reader Verification Plates
Fisherbrand
Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Have Questions?
Catalog NumberProduct Type
FKS11958995Multiskan Verification Plate, 8 wavelengths
Catalog number FKS11958995
Price (SGD)
16,005.00
EA
Add to cart
Product Type:
Multiskan Verification Plate, 8 wavelengths
Price (SGD)
16,005.00
EA
Add to cart
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation. 
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeMultiskan Verification Plate, 8 wavelengths
For Use With (Equipment)Multiskan FC
Plate TypeVerification Plate
Product LineMultiskan™
IncludesNPL (National Physical Laboratory) traceable Multiskan™ Verification Plate
Certificate of Calibration
USB memory device including SkanIt™ software sessions and reference files
Carrying case for Multiskan™ Verification Plate
Unit SizeEA