Borosilicate Glass Medium Wall Rimmed Test Tube
Borosilicate Glass Medium Wall Rimmed Test Tube
Actual product may vary
Fisherbrand
Borosilicate Glass Medium Wall Rimmed Test Tube
Manufactured from borosilicate glass with a medium wall design. Fisherbrand™ Borosilicate Glass Medium Wall Rimmed Test Tubes are ideal for chemical and biological work.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FKS1195219812 mm100 mm
Catalog number FKS11952198
Price (SGD)
-
Diameter (Metric):
12 mm
Length (Metric):
100 mm
  • Features: rimmed medium walls
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Length (Metric)100 mm
MaterialBorosilicate Glass
Quantity100 Pack
RimmedYes
Diameter (Metric)12 mm
Unit SizePK