▸
Pipette Products
▸
Pipette Tips
▸
Universal Pipette Tips
▸
Maxi Pipet Tips
Actual product may vary
Fisherbrand
Maxi Pipet Tips
For use with large volume pipets
Have Questions?
Change view
Catalog Number
Length (Metric)
FKS11947754
161 mm
FIS02-681-412
161 mm
FIS02-707-466
152 mm
FKS11957754
152 mm
4 Options
Catalog number
FKS11947754
Price (SGD)
168.00
Pack
Add to cart
Length (Metric):
161 mm
Price (SGD)
168.00
Pack
Add to cart
For use with large volume pipets
Designed for use when transferring large sample volume
Advanced molding technology ensure tips are straight
Alternative option to serological pipets
Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications
Volume (Metric)
1 to 10 mL
Sterility
Non-sterile
Color
Natural
Autoclavable
Autoclavable
Length (English)
6.44 in.
Length (Metric)
161 mm
Format
Bulk
Tip Style
Macro
For Use With (Equipment)
Gilson Pipetman P10
Material
Polypropylene