MBL Volumetrics™ Glass Graduated Cylinder
Actual product may vary
Fisherbrand
MBL Volumetrics™ Glass Graduated Cylinder
Manufactured from chemically resistant borosilicate glass. MBL Volumetrics™ Glass Graduated Cylinders are a reliable choice for measuring liquids and solutions in the laboratory.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS1194540610 mL0.2 mL
Catalog number FKS11945406
Price (SGD)
83.00
Pack
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (SGD)
83.00
Pack
Manufactured from chemically resistant borosilicate glass. MBL Volumetrics™ Glass Graduated Cylinders are a reliable choice for measuring liquids and solutions in the laboratory.
Features
  • Easy to read graduations in blue enamel
  • Sturdy hexagonal foot
  • Complies with ISO 4788
Specifications
Tolerance±0.2 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassClass B
Capacity (Metric)10 mL
For Use With (Application)Measuring Liquids and Solutions in the Laboratory
Height (Metric)140 mm
ScaleBlue enamel
Graduations0.2 mL
Base TypeHexagonal
Scale ColorBlue
ShapeRound
Product TypeGraduated Cylinder
Unit SizePack