▸
Microscopes
▸
Microscope Lamps and Accessories
▸
Microscope Accessories and Replacement Parts
▸
Immersion Oil for Microscopes
Actual product may vary
Fisherbrand
Immersion Oil for Microscopes
Use Leica™ Immersion Oil with microscope to increase the resolving power and aperture of the objective lens.
Have Questions?
Catalog Number
Refractive Index
FKS11944399
Type F
Catalog number
FKS11944399
Price (SGD)
-
Refractive Index:
Type F
10mL bottle
For Leica Microscopes
Specifications
Type
Immersion Oil
Packaging Type
1 Pack
Quantity
10 mL
Refractive Index
Type F
Size
10mL