Octahedral PTFE Stir Bar
Octahedral PTFE Stir Bar
Actual product may vary
Fisher Catalogue
Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
FKS1194357425 mm8 mm
FKS1192357415 mm8 mm
Catalog number FKS11943574
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
25 mm
Diameter (Metric):
8 mm
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Diameter (English)0.3125 in.
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Length (English)1 in.
Diameter (Metric)8 mm
Length (Metric)25 mm
Quantity5/Pk.
ShapeOctahedral
Unit SizePK