Graduated Plastic Beakers with Handle
Graduated Plastic Beakers with Handle
Actual product may vary
Fisherbrand
Graduated Plastic Beakers with Handle
Manufactured from polypropylene to be virtually indestructible and highly transparent. Buerkle™ Graduated Plastic Beakers with Handle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11941521500 mL
FKS118664332000 mL
Catalog number FKS11941521
Price (SGD)
26.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
26.00
EA
Add to cart
  • Permanent blue ink graduations 
  • Translucent
  • Autoclavable to 121°C 
  • Suitable for foodstuff
  • Integral handle and spout for easy drip-free pouring
Specifications
ColorTransparent
Diameter (Metric) Base160 mm
Diameter (Metric)Top210 mm
Capacity (Metric)500 mL
MaterialPolypropylene
For Use With (Application)Suitable for Foodstuff
Height (Metric)270 mm
IncludesHandle
Quantity6/Pk.
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
ScaleBlue Graduated Scale
Unit SizeEA