Check Order Status
Plastilab™ Tapered Straight Connectors
Actual product may vary
Fisherbrand
Plastilab™ Tapered Straight Connectors
For straight connection of tubing with different diameters.
Have Questions?
Catalog Number
Diameter (Metric)
Packaging Quantity
FKS11934349
6-8 mm
10/Pk.
Catalog number
FKS11934349
Price (SGD)
26.00
PK
Add to cart
Diameter (Metric):
6-8 mm
Packaging Quantity:
10/Pk.
Price (SGD)
26.00
PK
Add to cart
For straight connection of tubings with different diameters.
Constructed from polypropylene
Autoclavable
Specifications
Diameter (Metric)
6-8 mm
Autoclavable
Autoclavable
Material
Polypropylene
For Use With (Application)
For straight connection of different diameter tubing
Packaging Quantity
10/Pk.
Port Size
3.5mm bore
Shape
Straight
Product Type
Straight Tapered Tubing Connector