Check Order Status
Soda Lime Glass Durham Test Tubes
Actual product may vary
Fisherbrand
Soda Lime Glass Durham Test Tubes
Detects the production of gas by microorganisms. Fisherbrand™ soda lime glass durham test tubes allow visualization of trapped air bubbles.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11932218
50 mm
7.5 mm
FKS11912218
30 mm
6.5 mm
2 Options
Catalog number
FKS11932218
Price (SGD)
-
Length (Metric):
50 mm
Diameter (Metric):
7.5 mm
Used in microbiology
Fill durham with solution and insert upside down into a second tube
Rimless tube facilitates execution
Wall Thickness: 0.65mm
Specifications
Length (Metric)
50 mm
Material
Soda Lime Glass
For Use With
used in microbiology to detect production of gas by microorganisms
Quantity
1000 Pack
Rimmed
No
Diameter (Metric)
7.5 mm
Product Type
Durham Tube