Check Order Status
Polypropylene Graduated Container Lids
Actual product may vary
Fisherbrand
Polypropylene Graduated Container Lids
For polypropylene graduated containers 14-955-101,14-955-103, 14-955-104 and 14-955-106
Have Questions?
Catalog Number
Sterility
FKS11924395
Sterile
Catalog number
FKS11924395
Price (SGD)
103.00
PK
Add to cart
Sterility:
Sterile
Price (SGD)
103.00
PK
Add to cart
For use with the following Fisherbrand Specimen Containers:
14-955-101, -103 (4.5 oz.)
14-955-104, -106 (8 oz.)
Specifications
Product Type
Lid
Material
Polyethylene
Sterility
Sterile
Autoclavable
Not Autoclavable
For Use With (Equipment)
Polypropylene graduated containers
No. per Pack
500/Pk.