NucleoBond™ Xtra Combi Rack
NucleoBond™ Xtra Combi Rack
Actual product may vary
NucleoBond™ Xtra Combi Rack
NucleoBond™ Xtra Combi Rack
Fisherbrand
NucleoBond™ Xtra Combi Rack
Rack for use with NucleoBond Xtra Midi / Maxi, Xtra BAC, AX 100, AX 500, AX 2000, and AX 10000 Columns.
Have Questions?
Catalog NumberQuantity
FKS119225721 Ea.
Catalog number FKS11922572
Price (SGD)
-
Quantity:
1 Ea.
NucleoBond™ Xtra is a new generation of anion exchangers featuring fast filtration, high binding capacity and high flow rate. They yield ultra-pure plasmid DNA in about half the time compared with other Midi or Maxi kits based on anion-exchange chromatography. NucleoBond™ Xtra Midi and Maxi kits contain enlarged columns creating shorter silica resin beds.
  • Holds up to six (6) NucleoBond™ Xtra Midi and four (4) NucleoBond™ Xtra Maxi Columns
The Xtra Combi Rack holds columns from either kit; it is sold separately.
Specifications
DescriptionHolds up to six (6) NucleoBond™ Xtra Midi and four (4) NucleoBond™ Xtra Maxi Columns
Final Product TypeNucleo Bond Xtra Combi Rack
For Use With (Equipment)NucleoBond Xtra Midi, Maxi, BAC, AX 100, 500, 2000, And 10000, BAC 100 Columns
Product TypeNucleo Bond Xtra Combi Rack
Quantity1 Ea.
Unit SizeEA
Contents & Storage
1 Ea., 15°C to 25°C