For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Type
FKS11922355
0.1 to 0.5 mL
Filter Card
FIS5991022
0.1 to 0.5 mL
Filter Card
2 Options
Catalog number FKS11922355
Price (SGD)
264.00
PK
Add to cart
Volume (Metric):
0.1 to 0.5 mL
Type:
Filter Card
Price (SGD)
264.00
PK
Add to cart
TPX Single Sample Chamber
Reusable
Autoclavable
Filter Cards
Highly absorbent disposable filter cards for use with TPX chamber
Not for use with Cytofunnel™ disposable sample chambers