Nessler Cylinder
Nessler Cylinder
Actual product may vary
Fisherbrand
Nessler Cylinder
Manufactured from soda lime glass. Fisherbrand™ Nessler Cylinders are ideal for colorimetric analysis.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1192216850 mL
Catalog number FKS11922168
Price (SGD)
-
Capacity (Metric):
50 mL
  • For comparison of color and turbidity between liquids or solutions
  • Fixed values of 50mL or 100mL with 50mL graduation
  • Optically plane bottom provides maximum clarity for visual comparisons
  • Supplied in matched pairs to ensure uniformity in product for accuracy
Specifications
Base TypeFlat
Tolerance±0.4 mL
MaterialGlass
Diameter (Metric) Outside26 mm
Capacity (Metric)50 mL
Height (Metric)150 mm
Product TypeNessler Tube
Unit SizePK