Aluminum Weighing Dishes with Fluted Sides
Aluminum Weighing Dishes with Fluted Sides
Actual product may vary
Fisherbrand
Aluminum Weighing Dishes with Fluted Sides
Designed for moisture determination balances. Fisherbrand™ Aluminum Weighing and Drying Dish is ideal for general purpose weighing, storage, processing or drying. Extra large, light-weight aluminum dishes measure 108mm diameter. Includes 50 disposable, stackable dishes per pack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeDiameter (Metric)Height (Metric)
FKS11918094Large70 mm16 mm
FKS11908094Medium57 mm12 mm
Catalog number FKS11918094
Price (SGD)
44.00
PK
Add to cart
Size:
Large
Diameter (Metric):
70 mm
Height (Metric):
16 mm
Price (SGD)
44.00
PK
Add to cart
  • Ideal for sample weighing, dispensing, storage, food processing, evaporating or drying
  • Also ideal as a dust cover for open vessels or as sample holders
  • Made from oil-free, light gauge aluminum that is FDA/food-grade compliant
  • Tapered walls promote stable stacking
  • Available in 4 varieties
    • Boats: 3 sizes with crimped sides, flat bottoms and finger tabs that can be etched for easy identification (100/pack)
    • Dish: 1 large size, designed for moisture determination balance (50/pack)
Specifications
Bottom ShapeFlat
Capacity (Metric)50 mL
DisposableSingle-use
MaterialAluminum
ShapeRound
TypeWeighing Dish
For Use With (Application)For general purpose weighing, dispensing, storage, food processing, evaporating, or drying
SizeLarge
Height (Metric)16 mm
Diameter (Metric)70 mm
Unit SizePK