Premium Microcentrifuge Tubes: 0.6mL
Premium Microcentrifuge Tubes: 0.6mL
Actual product may vary
Fisher Catalogue
Premium Microcentrifuge Tubes: 0.6mL
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS11916955Natural
FIS05-408-121Red
FIS05-408-123Blue
FIS05-408-120Natural
FIS05-408-122Yellow
FIS05-408-125Amber
FIS05-408-128Assorted
Catalog number FKS11916955
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Capacity: 0.6mL
  • High-clarity polypropylene
  • Graduated in 0.1mL markings
  • Withstand forces up to 30,000 × g
  • Easy-open caps with needle insertion spot and textured marking area
  • Labeling area on tube exterior
  • Highly polished interior for maximum sample recovery
  • Temperature range of -80° to +121°C
  • Fit all standard rotors
  • Nonpyrogenic
  • Autoclavable at 121°C/15psi for 15 minutes
Specifications
Cap StyleSnap Cap with Flat Top
Certifications/ComplianceRNase, DNase-free
AutoclavableAutoclavable
Capacity (Metric)0.5 mL (0.6 mL max.)
ColorNatural
Length (Metric)30.5 mm
MaterialPolypropylene
IncludesNeedle insertion spot
Label TypeLabeling Area
GraduatedYes
Closure TypeFlat Snap Cap
Max. RCF30,000 x g
Diameter (Metric)7.5 mm
Unit SizePK