11 mm Glass Crimp Top Vials
11 mm Glass Crimp Top Vials
Actual product may vary
Fisherbrand
11 mm Glass Crimp Top Vials
Use these 11 mm clear glass vials with snap caps or aluminum crimp seal closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)TypeBase StyleMarking Spot
FKS119157132 mLCrimp Top VialFlat BottomNo
FIS03-375-27A2 mLSnap-It VialFlat BottomYes
FIS10-800-105300 μLSnap Cap VialFused ConicalYes
FIS03-377-9992 mLCrimp Top VialFlat BottomYes
Catalog number FKS11915713
Price (SGD)
72.00
PK
Add to cart
Volume (Metric):
2 mL
Type:
Crimp Top Vial
Base Style:
Flat Bottom
Marking Spot:
No
Price (SGD)
72.00
PK
Add to cart
Use Thermo Scientific™ 11 mm Glass Crimp Top Vials with aluminum crimp seal closures. These high-quality 11 mm glass vials are 2 mL, 12 x 32 mm and fit most brands of autosamplers. Made of superior quality 33 expansion borosilicate clear (Type 1, Class A), they meet all requirements of US, EU, JPN Pharmacopia. Choose from Thermo Scientific™, Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
Choose the right vial for your needs
  • Easy-to-fill wide neck opening vials require 6 mm diameter micro-inserts
  • Identify sample and volume with graduated, write-on patch provided on certain vials
  • Get maximum sample extraction without need for separate inserts by using Microsampling and High Recovery Vials
  • Obtain optimal recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional)
Select the insert for your sample
  • Polyspring inserts are self-aligning and provide a cushion against needle contact
  • Precision point inserts minimize residual sample loss
  • Pulled point inserts are an economical choice for noncritical applications
Autosampler compatibility*
Compatible with most HPLC and GC autosamplers
*See the Columns and Consumables Catalog for list of autosampler compatibility.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTarget LoVial, 2mL Clear Glass Vial
Diameter (Metric) Opening11 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2 mL
Volume (Metric) Residual<170μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Closure TypeCrimp Cap
Height (Metric)32 mm
Quantity100
TypeCrimp Top Vial
Closure Size11 mm
Marking SpotNo
Volume (Metric) Usable1.5 mL
Unit SizePK