Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size | Height (Metric) | Tolerance |
---|---|---|---|---|
FKS11911528 | 250 mL | 14/23 | 220 mm | ±0.12 mL |
FKS10059279 | 2000 mL | 29/32 | 370 mm | ±0.5 mL |
FKS11363634 | 2 mL | 7/16 | 65 mm | ±0.02 mL |
FKS11373634 | 5 mL | 10/19 | 70 mm | ±0.02 mL |
FKS11393634 | 20 mL | 10/19 | 110 mm | ±0.03 mL |
FKS11303644 | 25 mL | 10/19 | 110 mm | ±0.03 mL |
FKS11313644 | 50 mL | 12/21 | 140 mm | ±0.05 mL |
FKS11323644 | 100 mL | 14/23 | 170 mm | ±0.08 mL |
FKS11333644 | 200 mL | 14/23 | 210 mm | ±0.1 mL |
FKS11353644 | 500 mL | 19/26 | 260 mm | ±0.2 mL |