Whatman™ Drying Rack for 903 Protein Saver Cards
Whatman™ Drying Rack for 903 Protein Saver Cards
Actual product may vary
Fisherbrand
Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
Have Questions?
Catalog NumberIncludes
FKS11909134Hook-and-Loop Mounting Sticker
Catalog number FKS11909134
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Includes:
Hook-and-Loop Mounting Sticker
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
  • For use with neonatal human diagnostic testing cards
  • CE Marked
  • Accommodate multiple collection forms at one time
Specifications
IncludesHook-and-Loop Mounting Sticker
Product TypeDrying Rack
For Use With (Equipment)903 Protein Saver Card
Quantity10 Pack
Unit SizePK