Acrylic 3-Compartment Dispensing Bin
Acrylic 3-Compartment Dispensing Bin
Actual product may vary
Fisherbrand
Acrylic 3-Compartment Dispensing Bin
Keep spare lab and office items easily accessible. Acrylic Dispensing Bins have large openings with hinged lids for easy filling and dispensing. The sturdy clear bins make it easy to monitor inventory. Four compartments help organize small parts.
Have Questions?
Catalog NumberColor
FKS11907994Transparent
Catalog number FKS11907994
Price (SGD)
419.00
EA
Add to cart
Color:
Transparent
Price (SGD)
419.00
EA
Add to cart
  • Ideal for pipette tips, tubes, fittings, syringe filters, septa and other small lab and office items
  • Made from 3 mm thick, transparent acrylic for at-a-glance inventory monitoring
  • Hinged lids over generous-sized openings for rapid filling and dispensing
  • Three compartments help to organize and categorize items
  • Flip-top covers keep products secure and dust free
  • Includes non-skid rubber feet
Specifications
TypeStorage Bin
MaterialAcrylic
ColorTransparent
ShapeRectangular
For Use With (Application)To keep spare parts, vials, syringe filters, pipettor tips, septa and other small laboratory and office items
Quantity1 Ea.
Length (English)8.3 in.
Length (Metric)210 mm
Width (Metric)155 mm
Height (Metric)212 mm
Unit SizeEA