Check Order Status
96-Well Polypropylene Microplates
Actual product may vary
Fisher Catalogue
96-Well Polypropylene Microplates
Ideal for storage—feature generous 500μL well volumes
Have Questions?
Catalog Number
Color
FKS11907954
Natural
Catalog number
FKS11907954
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
Polypropylene
Minimum binding
Resistant to many reagents
Withstand temperatures from -80° to +121°C (-112° to +250°F)
Automation-friendly design optimized for HTS
Specifications
Format
Standard
Array
8 x 12
Color
Natural
Material
Polypropylene
No. of Wells
96
Sterility
Non-sterile
Well Shape
Round
For Use With (Application)
For storage feature generous 500μL well volumes