Low-Retention Microcentrifuge Tubes
Low-Retention Microcentrifuge Tubes
Actual product may vary
Low-Retention Microcentrifuge Tubes
Provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityCapacity (Metric)Closure Type
FKS11906965Non-sterile2 mLSnap Cap
FIS02-681-311Non-sterile0.6 mLSnap Cap
FIS02-681-320Non-sterile1.5 mLSnap Cap
FIS02-681-321Non-sterile2 mLSnap Cap
FIS02-681-331Sterile1.5 mLAttached Cap
FIS02-681-332Sterile2 mLAttached Cap
Catalog number FKS11906965
Price (SGD)
29.00
PK
Add to cart
Sterility:
Non-sterile
Capacity (Metric):
2 mL
Closure Type:
Snap Cap
Price (SGD)
29.00
PK
Add to cart
Snap-Cap™ lid provides a secure seal—even over extended periods. Critical applications where minute sample recovery is important.
  • Manufactured from durable chemical-resistant polypropylene
  • 0.6mL tube is graduated in 0.2mL increments; 1.5mL tube, in 0.5mL increments with an additional graduation at 0.1 and 2.0mL size, in 0.5mL increments
  • Designed for use in critical applications where minute sample recovery is important
  • Withstand speeds up to 30,000xG
  • Recommended for freezer-to-centrifuge use
  • Autoclavable up to 120°C and withstand freezing temperatures as low as -80°C
  • RNase/DNase free
Specifications
Cap StyleSnap Cap
AutoclavableAutoclavable
Capacity (Metric)2 mL
ColorNatural
Length (Metric)40 mm
MaterialPolypropylene
For Use With (Application)Critical applications where minute sample recovery is important
PackagingBulk
SterilityNon-sterile
GraduatedYes
Closure TypeSnap Cap
Max. RCF30,000 x g
Diameter (Metric)13 mm
Unit SizePK