20 mm Headspace Vials, Septum, and Caps
20 mm Headspace Vials, Septum, and Caps
Actual product may vary
Fisherbrand
20 mm Headspace Vials, Septum, and Caps
Headspace vials are available in clear or amber glass with a round or flat base, a crimp seal with a beveled or square edge finish, or with a screw-thread finish. Headspace vials, septum, and caps can be purchased in various quantities separately or together as convenience kits to fit your laboratory needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorTypeDiameter (Metric) InnerMaterial
FKS11905713Clear20 mm Headspace Crimp Vial23 mmGlass
FIS03-377-119Clear20 mm Headspace Crimp Vial23 mmGlass
FIS03-377-118Clear20 mm Headspace Crimp Vial Convenience Kit20 mmGlass
FIS03-377-123Gray, Silver20 mmPTFE, Butyl Rubber, Aluminum-PR
FIS03-375-22ESilver, Tan, WhiteCrimp CapPTFE, Silicone, Aluminum
FIS03-397-51Clear, Blue Translucent, Gold20 mmPTFE, Silicone, Tinplate
Catalog number FKS11905713
Price (SGD)
207.00
PK
Add to cart
Color:
Clear
Type:
20 mm Headspace Crimp Vial
Diameter (Metric) Inner:
23 mm
Material:
Glass
Price (SGD)
207.00
PK
Add to cart
Thermo Scientific™ headspace vials are available in clear or amber glass with a round or flat base, a crimp seal with a beveled or square edge finish, or with a screw-thread finish. Headspace vials, septum, and caps can be purchased in various quantities separately or together as convenience kits to fit your laboratory needs.
  • Type 1 borosilicate glass
  • Clear glass vials with 20mm crimp seal finish are designed to fit most headspace autosamplers
  • Flat bottom vials maximize heating efficiency when used with bottom
  • Round bottom vials are more easily handled by robotic arms that lift the vial from the tray
  • Round bottom vials distribute the internal pressure created at high temperatures across the glass surface
  • Beveled edge finish features a sturdy rim that presses into the septum for a more effective seal
Compatibility
  • Flat bottom vials feature a longer neck for fit with Carlo Erba™ and Agilent™ units
  • Round bottom vials are designed for Perkin Elmer™, Tekmar™ and Varian™ units
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description20mL; Flat Bottom; Beveled Edge Crimp Top. Contains Vials only. Caps needed: 03-375-22E.
Diameter (Metric) Opening20 mm
Diameter (Metric) Inner23 mm
ColorClear
Diameter (Metric) Outer23 mm
MaterialGlass
Volume (Metric)21.5 mL
Base StyleFlat Bottom
Volume (Metric) Total21.5 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Quantity100
Type20 mm Headspace Crimp Vial
Closure Size20 mm
Marking SpotNo
Volume (Metric) Usable20 mL
For Use With (Equipment)Carlo Erba™ and Agilent™ Units
Unit SizePK