Soda Lime Glass Medium Wall Rimless Test Tube
Soda Lime Glass Medium Wall Rimless Test Tube
Actual product may vary
Fisherbrand
Soda Lime Glass Medium Wall Rimless Test Tube
Manufactured from soda lime glass and constructed with medium walls. Fisherbrand™ Soda Lime Glass Medium Wall Rimless Test Tubes are ideal for general mixing and laboratory work.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FKS1190219816 mm125 mm
Catalog number FKS11902198
Price (SGD)
-
Diameter (Metric):
16 mm
Length (Metric):
125 mm
  • Features: rimless medium walls
  • Use for: bacteriological performance
  • Complies to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Length (Metric)125 mm
MaterialSoda Lime Glass
Quantity100 Pack
RimmedNo
Diameter (Metric)16 mm
Unit SizePK