1x1 image pixel for data collection
Transparent Polypropylene Powder Funnel
Transparent Polypropylene Powder Funnel
Actual product may vary
Fisherbrand
Transparent Polypropylene Powder Funnel
Use these funnels for a variety of bottling and decanting applications in the food industry. VITLAB™ Transparent Polypropylene Powder Funnels feature a short, wide stem. Ideal for transfer of powdered and granular substances.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)
FKS11897890110 mm
FKS1071353368 mm
FKS1187789075 mm
Catalog number FKS11897890
Price (SGD)
135.00
PK
Add to cart
Height (Metric):
110 mm
Price (SGD)
135.00
PK
Add to cart
  • Transparent polypropylene funnel
  • Rapid flow due to a steep 60° angle
Specifications
Diameter (Metric)Top120mm
Diameter (Metric) Stem26mm
MaterialPolypropylene
Height (Metric)110 mm
Angle60°
AutoclavableAutoclavable
For Use With (Application)Suitable for food contact
Length (Metric) Stem20mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link