Sterilin™ Clear Microtiter™ Plates
Sterilin™ Clear Microtiter™ Plates
Actual product may vary
Fisherbrand
Sterilin™ Clear Microtiter™ Plates
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapeSterility
FKS11897403Conical Bottom (V-well)Non-sterile
FKS11349163Flat Bottom (F-well)Non-sterile
FKS11399153Round Bottom (U-well)Sterile
FKS11319163Flat Bottom (F-well)Sterile
FKS11329163Not ApplicableSterile
Catalog number FKS11897403
Price (SGD)
261.00
PK
Add to cart
Well Shape:
Conical Bottom (V-well)
Sterility:
Non-sterile
Price (SGD)
261.00
PK
Add to cart
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.
Features
  • Manufactured from virgin polystyrene for excellent optical clarity
  • Manufactured in ISO Class 7 Clean Room without surface treatment for a medium binding surface
  • Available irradiated for sterility and a more hydrophilic surface, providing high binding
  • Suitable for use at a wavelength of 340nm
  • Minimizes the risk of contamination from surrounding wells with condensation rings on the lid raised well rims on the base.
  • Ensures even temperature distribution around each well with a hanging well design
Easy Sample Identification
  • Frosted writing area on the end wall
  • Orientation corners
  • Alphanumeric labeling
Note:
Plates are supplied without a lid (can be purchased separately).
Ordering Information:
Product is not available in all geographic regions.
Specifications
Certifications/ComplianceCE marked in accordance with the European Directive 98/72/EC
FormatStandard
Array8 x 12
Packaging50 Cases
ColorClear
Product TypeMicrotiter Plates
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
No. per Case50
No. per Pack5
Quantity50/Pk.
Unit SizePK