HDPE Sealable Wide Mouth Bottles
HDPE Sealable Wide Mouth Bottles
Actual product may vary
Fisherbrand
HDPE Sealable Wide Mouth Bottles
Sturdy bottle with sealing eyelets on the screw cap and the bottle. Particularly suited for sending samples. Seals provide increased sample authenticity. Easy to fill due to wide opening. Particularly suitable for viscous media, powder, granules and pastes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS118927311000 mL
FKS11882731500 mL
Catalog number FKS11892731
Price (SGD)
35.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
35.00
PK
Add to cart
  • HDPE
  • Leakproof
  • Easy to fill due to wide-necked opening
SampleSafe seals available as accessories
Specifications
Capacity (Metric)1000 mL
Diameter (Metric) Inner Neck50 mm
MaterialHDPE
Closure IncludedYes
MouthWide
Height (Metric)186 mm
Diameter (Metric)100 mm
Closure TypeScrew Cap
Unit SizePK