Desiccator Cabinet
Desiccator Cabinet
Actual product may vary
Fisherbrand
Desiccator Cabinet
Desiccator cabinets can be used in refrigerators or at room temperature for dry storage of moisture-sensitive materials.
Have Questions?
Catalog NumberHeight (Metric)
FKS1189272221.3 cm
Catalog number FKS11892722
Price (SGD)
1,131.00
EA
Add to cart
Height (Metric):
21.3 cm
Price (SGD)
1,131.00
EA
Add to cart
  • Large doors maximize access to interior space
  • Perforated shelves facilitate circulation of dry air
Specifications
ColorBlue
Cabinet StyleVertical
MaterialCopolyester
No. of Shelves1
No. per Pack1 Each
Shelf PositionsFixed
Shelf TypePerforated
Width (English)13.4 in.
Width (English) Door13.4 in.
Width (Metric)34.1 cm
Width (Metric) Door34.1 cm
Height (English)8.4 in.
Height (Metric)21.3 cm
Depth (English)16.3 in.
Depth (Metric)41.4 cm
Unit SizeEA