BioLite™ Cell Culture Treated Flasks
BioLite™ Cell Culture Treated Flasks
Actual product may vary
Fisherbrand
BioLite™ Cell Culture Treated Flasks
Thermo Scientific™ BioLite™ Cell Culture Treated Flasks featuring standard cell culture-treated surface, are a limited line of cell culture vessels for a wide range of cell types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaClosure Type
FKS1188423575 cm2Vented
FKS1183076525 cm2Vented
FIS12-556-01075 cm2Vented
Catalog number FKS11884235
Price (SGD)
390.00
PK
Add to cart
Culture Area:
75 cm2
Closure Type:
Vented
Price (SGD)
390.00
PK
Add to cart
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated flasks with filter cap and seal cap options suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Sterile per ISO 11137, SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• BSE/TSE Free
• RNase/DNase Free
• Human DNA Free
• USP Class VI raw material
• Shelf life: 2 years
Specifications
No. per Case100
No. per Pack5
Product TypeCell Culture Flask
Surface CoatingCell Culture Treated
SterilitySterile
Bottom ShapeFlat
Closure TypeVented
Culture Area75 cm2
Surface TreatmentCell Culture-treated
Volume (Metric) Working25 mL
Unit SizePK