Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders
Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders
Actual product may vary
Fisherbrand
Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders
Constructed of borosilicate glass and complies with DIN EN ISO 4788 standards. BRAND™ Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders include integral spout and hexagonal glass base for stability. DE-M Marking amber stain graduations provide quick and easy volume assessment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS1188399010 mL0.2 mL
FKS11824000500 mL5 mL
FKS1180400050 mL1 mL
Catalog number FKS11883990
Price (SGD)
82.00
PK
Add to cart
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (SGD)
82.00
PK
Add to cart
  • Class A tall form measuing cylinder constructed of borosilicate glass
  • Complies with DIN EN ISO 4788 standards
  • Autoclavable
  • Calibrated to contain (TC, In).
  • With integral spout and hexagonal glass base for stability
  • DE-M Marking amber stain graduations provide quick and easy volume assessment
  • USP individual certificate or DAkkS calibration certificate available upon request
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorAmber
ShapeRound
Certifications/ComplianceDIN EN ISO 4788
ClassA
Height (Metric)140 mm
Tolerance Metric±0.1 mL
TypeGraduated Measuring Cylinder
Item DescriptionDE-M Marking, amber stain graduations
MaterialBorosilicate Glass
Capacity (Metric)10 mL
IncludesBatch Certificate
Graduations0.2 mL
AutoclavableAutoclavable
Subdivision0.2mL
Unit SizePK