Translucent Silicone Tubing
Translucent Silicone Tubing
Actual product may vary
Translucent Silicone Tubing
Translucent Silicone Tubing
Fisherbrand
Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterOverall Length (Metric)
FKS118762934 mm10 mm5 m
FKS1171385510 mm14 mm5 m
FKS117638453 mm6 mm5 m
FKS117634065 mm8 mm25 m
Catalog number FKS11876293
Price (SGD)
69.00
EA
Add to cart
Diameter (Metric) Inner:
4 mm
Diameter (Metric) Outer:
10 mm
Overall Length (Metric):
5 m
Price (SGD)
69.00
EA
Add to cart
  • Translucent silicone tubing
  • FDA compliant (food)
Specifications
Certifications/ComplianceFDA, BFR and WRAS approved
AutoclavableAutoclavable
Diameter (Metric) Inner4 mm
MaterialSilicone
Diameter (Metric) Outer10 mm
Overall Length (Metric)5 m
Thickness (Metric)3 mm
For Use WithPeristaltic pumps
For Use With (Application)Pharmaceutical , biotechnology, transfer sterile fluids
Min. Operating Temperature (Metric)-40°C
SterilitySterile
ColorTranslucent
FlexibilityFlexible
Max. Operating Temperature (Metric)250°C
Unit SizeEA