Check Order Status
Conical Containers
Actual product may vary
Conical Containers
Provides safe and secure storage in the laboratory. Gosselin™ Conical Containers are manufactured from either clear or white polypropylene.
Have Questions?
Catalog Number
Capacity (Metric)
Color
Diameter (Metric)
Closure Included
FKS11871091
200 mL
Natural
62 mm
Yes (Unassembled)
Catalog number
FKS11871091
Price (SGD)
655.00
PK
Add to cart
Capacity (Metric):
200 mL
Color:
Natural
Diameter (Metric):
62 mm
Closure Included:
Yes (Unassembled)
Price (SGD)
655.00
PK
Add to cart
Shock resistant
Molded graduations outside on clear containers; molded internal graduations on white containers
Specifications
Product Type
Container
Closure Included
Yes (Unassembled)
Graduated
Yes
Material
Polypropylene
Shape
Conical, Cylindrical
Sterility
Non-sterile
Capacity (Metric)
200 mL
Certifications/Compliance
EC N°1935/2004
Closure Color
Natural
Closure Type
Snap Cap
Color
Natural
Diameter (Metric)
62 mm
Graduations
25 mL
Height (Metric)
88 mm
Includes
Natural LDPE snap cap
No. per Case
660/Cs.