Stainless Steel Ice Borer
Stainless Steel Ice Borer
Actual product may vary
Stainless Steel Ice Borer
Stainless Steel Ice Borer
Fisherbrand
Stainless Steel Ice Borer
Stainless steel ice borer is a sampler for deep-frozen and semi-solid substances products in the food industry.
Have Questions?
Catalog NumberDiameter (Metric) Outer Top
FKS1186639321 mm
Catalog number FKS11866393
Price (SGD)
1,719.00
PK
Add to cart
Diameter (Metric) Outer Top:
21 mm
Price (SGD)
1,719.00
PK
Add to cart
  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilisable
  • 20 cm boring depth
  • The sharp sawing crown of the very robust hollow borer rapidly cuts into the sample
  • A cylindrical sample (cross-sectional sample) of up to 20 cm can be cut out
Specifications
Product TypeIce Borer
MaterialStainless Steel (V4A)
AutoclavableAutoclavable
DescriptionI.D. 16 mm
For Use With (Application)Ideal for Deep-Frozen and Semi-Solid Substances Products in The Food Industry
Diameter (Metric) Outer Top21 mm
Length (Metric)200 mm
Unit SizePK