Sub-Gel Midi Horizontal Gel Electrophoresis Unit
Sub-Gel Midi Horizontal Gel Electrophoresis Unit
Actual product may vary
Fisherbrand
Sub-Gel Midi Horizontal Gel Electrophoresis Unit
For rapid electrophoresis and quick checking of samples from PCR and cloning.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS11853303220 x 125 x 90 mm
Catalog number FKS11853303
Price (SGD)
1,514.00
EA
Add to cart
Dimensions (L x W x H):
220 x 125 x 90 mm
Price (SGD)
1,514.00
EA
Add to cart
  • Runs up to 100 samples
  • Low buffer volumes
  • Injection-moulded construction
Specifications
TypeSub-Gel Midi Horizontal Gel Electrophoresis Unit
Gel Dimensions70L x 100 mm W, 100L x 100 mm W
Sample Size100
Volume (Metric) Buffer300 mL
Dimensions (L x W x H)220 x 125 x 90 mm
Unit SizeEA