Silberbrand™ ETERNA Class B Borosillicate Glass Low Form Measuring Cylinder
Silberbrand™ ETERNA Class B Borosillicate Glass Low Form Measuring Cylinder
Actual product may vary
Fisherbrand
Silberbrand™ ETERNA Class B Borosillicate Glass Low Form Measuring Cylinder
Manufactured from DURAN™ Borosilicate Glass and complies with DIN EN ISO 4788 standards. BRAND™ Silberbrand™ ETERNA Class B Borosilicate Glass Low Form Measuring Cylinder with amber stain graduations provide quick and easy volume assessment with opaque, light-colored liquids. Includes integral spout to prevent spillage while pouring while hexagonal glass base provides stability.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS1183837110 mL1 mL
Catalog number FKS11838371
Price (SGD)
66.00
PK
Add to cart
Capacity (Metric):
10 mL
Graduations:
1 mL
Price (SGD)
66.00
PK
Add to cart
  • Manufactured from DURAN™ Borosilicate Glass
  • Complies with DIN EN ISO 4788 standards
  • Amber stain graduations provide quick and easy volume assessment with opaque, light-colored liquids.
  • Includes integral spout to prevent spillage while pouring
  • Hexagonal glass base provides stability
Specifications
Base TypeHexagonal
CalibrationTo Contain
ShapeRound
Certifications/ComplianceDIN EN ISO 4788
ClassB
Height (Metric)90 mm
Tolerance Metric±0.3 mL
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Capacity (Metric)10 mL
Graduations1 mL
AutoclavableAutoclavable
Limit of Error±0.3mL
Subdivision1mL
Unit SizePK