Polypropylene Cylindrical Y Connectors
Polypropylene Cylindrical Y Connectors
Actual product may vary
Fisherbrand
Polypropylene Cylindrical Y Connectors
Safeguard a flexible and safe connection for demanding applications with these Y connectors. Buerkle™ Polypropylene Cylindrical Y Connectors are available in a variety of sizes to extend, connect, branch, join and repair hoses and tubing.
Have Questions?
Catalog NumberFor Use With
FKS118365837 to 9 mm Hose
Catalog number FKS11836583
Price (SGD)
60.00
PK
Add to cart
For Use With:
7 to 9 mm Hose
Price (SGD)
60.00
PK
Add to cart
  • Cylindrical Y hose connector
  • Available in a variety of sizes from 3 to 15mm dia.
  • Polypropylene
  • Temperature resistant up to +135°C max.
  • Sterilizable
Specifications
Certifications/ComplianceEU Foodstuffs, RoHS
AutoclavableAutoclavable
Diameter (Metric) Inner6 mm
MaterialPolypropylene
For Use With7 to 9 mm Hose
Packaging Quantity10/Pk.
Resistance PropertiesUp to 135°C Temperature-Resistant
ShapeCylindrical Y
ColorTransparent
Max. Operating Temperature (Metric)135°C
Product TypeY Connector
Unit SizePK