Check Order Status
Stainless Steel Compression Fittings
Actual product may vary
Fisherbrand
Stainless Steel Compression Fittings
Have Questions?
Catalog Number
For Use With
FKS11835453
0.125 in. NPT(M)
Catalog number
FKS11835453
Price (SGD)
156.00
EA
Add to cart
For Use With:
0.125 in. NPT(M)
Price (SGD)
156.00
EA
Add to cart
Specifications
Autoclavable
Autoclavable
Material
Stainless Steel
For Use With
0.125 in. NPT(M)
Min. Operating Temperature (Metric)
-40°C
Packaging Quantity
1 Pack
Max. Operating Temperature (Metric)
93°C
Product Type
Compression Fitting