Check Order Status
18/10 Stainless Steel Micro Spoon Spatulas, Spoon Shape
Actual product may vary
Fisher Catalogue
18/10 Stainless Steel Micro Spoon Spatulas, Spoon Shape
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Micro Spoon Spatulas comes in two types.
Have Questions?
Change view
Catalog Number
Length (Metric)
Product Type
Diameter (Metric) Outer
FKS11835071
185 mm
Micro Spoon Spatula
2mm
FKS10557122
210 mm
Spoon Spatula
4mm
2 Options
Catalog number
FKS11835071
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
185 mm
Product Type:
Micro Spoon Spatula
Diameter (Metric) Outer:
2mm
18/10 stainless steel
Specifications
Quantity
10 Pk.
Shape
Spoon End, Blade End
Product Type
Micro Spoon Spatula
Width (Metric)
5 mm
Width (Metric) Blade
5 mm
Diameter (Metric) Outer
2mm
Dimensions (L x W)
185 x 5 mm
Material
Stainless Steel (18/10)
Length (Metric)
185 mm