Adapters for Rectangular Buckets in A-4-81 Rotor
Adapters for Rectangular Buckets in A-4-81 Rotor
Actual product may vary
Adapters for Rectangular Buckets in A-4-81 Rotor
Adapters for Rectangular Buckets in A-4-81 Rotor
Fisherbrand
Adapters for Rectangular Buckets in A-4-81 Rotor
Add flexibility to A-4-81 Rotor for Eppendorf Centrifuge 5810/5810 R with these adapters
Have Questions?
Catalog NumberCapacity
FKS11833981180 to 250 mL
Catalog number FKS11833981
Price (SGD)
1,221.00
PK
Add to cart
Capacity:
180 to 250 mL
Price (SGD)
1,221.00
PK
Add to cart
  • Rotor adapters
  • For rectangluar buckets
  • Available in a wide range of sizes to suit various applications
Specifications
TypeAdapter
Tube TypeStandard, Vacutainer, Falcon Tubes and Flasks
Capacity180 to 250 mL
For Use With (Equipment)500mL Rectangular Bucket for Rotor A-4-81
Bore Size (Metric)62 mm
No. of Places1
For Use With (Application)400mL rectangular bucket for use with standard, Vacutainer, Falcon tubes and flasks
No. per Pack2 Pack
Unit SizePK